missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Prostasin/Prss8 Antibody, R&D Systems™

Goat Polyclonal Antibody
£168.00 - £452.00
Specifications
| Antigen | Prostasin/Prss8 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18769207
![]() |
R&D Systems
AF2968-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18021272
|
R&D Systems
AF2968 |
100 μg |
£452.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prostasin/Prss8 Polyclonal specifically detects Prostasin/Prss8 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| Prostasin/Prss8 | |
| Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| CAP1, channel-activating protease 1, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.120, prostasin, protease, serine, 8, Prss8, Serine protease 8 | |
| PRSS8 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Q99L44 | |
| 5652 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Prostasin/Prss8 Ala30-Gln289 Accession # Q99L44 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Prostasin/Prss8 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity is observed with recombinant human Prostasin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title