missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Prostasin/Prss8 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2968-SP
This item is not returnable.
View return policy
Description
Prostasin/Prss8 Polyclonal specifically detects Prostasin/Prss8 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| Prostasin/Prss8 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAP1, channel-activating protease 1, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.120, prostasin, protease, serine, 8, Prss8, Serine protease 8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5652 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Q99L44 | |
| PRSS8 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Prostasin/Prss8 Ala30-Gln289 Accession # Q99L44 | |
| 25 μg | |
| Primary | |
| Detects mouse Prostasin/Prss8 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity is observed with recombinant human Prostasin. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction