missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Progranulin/PGRN Antibody, R&D Systems™

Rat Monoclonal Antibody
£167.00 - £492.00
Specifications
| Antigen | Progranulin/PGRN |
|---|---|
| Clone | 333708 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18750837
![]() |
R&D Systems
MAB2557-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18176061
|
R&D Systems
MAB2557 |
500 μg |
£492.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Progranulin/PGRN Monoclonal specifically detects Progranulin/PGRN in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Progranulin/PGRN | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Acrogranin, CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN, Proepithelin | |
| Grn | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 333708 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P28798 | |
| 2896 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Progranulin/PGRN Thr18-Leu589 Accession # P28798 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects mouse Progranulin/PGRN in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human Progranulin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title