missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Progranulin/PGRN Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2557-SP
This item is not returnable.
View return policy
Description
Progranulin/PGRN Monoclonal specifically detects Progranulin/PGRN in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| Progranulin/PGRN | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Acrogranin, CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN, Proepithelin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2896 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 333708 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P28798 | |
| Grn | |
| Mouse myeloma cell line NS0-derived recombinant mouse Progranulin/PGRN Thr18-Leu589 Accession # P28798 | |
| 25 μg | |
| Primary | |
| Detects mouse Progranulin/PGRN in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human Progranulin is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction