missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Notch-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
£187.00 - £514.00
Specifications
| Antigen | Notch-3 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18725757
![]() |
R&D Systems
AF1308-SP |
25 μg |
£187.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18099641
![]() |
R&D Systems
AF1308 |
100 μg |
£514.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Notch-3 Polyclonal specifically detects Notch-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
| Notch-3 | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| Polyclonal | |
| Goat | |
| Mouse | |
| CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3, Notch3 | |
| NOTCH3 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF | |
| Unconjugated | |
| RUO | |
| Q61982 | |
| 4854 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Notch-3 Ala40-Glu468 Accession # Q61982 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Notch-3 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human Notch-3 is observed and less than 5% cross-reactivity with recombinant rat (rr) Notch-1 and rrNotch-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title