missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Notch-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF1308
This item is not returnable.
View return policy
Description
Notch-3 Polyclonal specifically detects Notch-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF-ready.
Specifications
| Notch-3 | |
| Polyclonal | |
| Unconjugated | |
| Q61982 | |
| NOTCH3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Notch-3 Ala40-Glu468 Accession # Q61982 | |
| 100 μg | |
| Primary | |
| Detects mouse Notch-3 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human Notch-3 is observed and less than 5% cross-reactivity with recombinant rat (rr) Notch-1 and rrNotch-2 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3, Notch3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4854 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction