missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Nope/IGDCC4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
£147.00 - £476.00
Specifications
| Antigen | Nope/IGDCC4 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18708127
![]() |
R&D Systems
AF1394-SP |
25 μg |
£147.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18002612
|
R&D Systems
AF1394 |
100 μg |
£476.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nope/IGDCC4 Polyclonal specifically detects Nope/IGDCC4 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| Nope/IGDCC4 | |
| Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| DDM36, hDDM36, IGDCC4, immunoglobulin superfamily DCC subclass member 4, immunoglobulin superfamily, DCC subclass, member 4, KIAA1628, likely ortholog of mouse neighbor of Punc E11, LOC57722, Neighbor of punc e11, NOPEFLJ42051, Protein DDM36 | |
| IGDCC4 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Q9EQS9 | |
| 57722 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Nope/IGDCC4, Gly22-His953, Accession # Q9EQS9 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Nope/IGDCC4 in ELISAs and Western blots. In sandwich immunoassays, less than 0.3% cross-reactivity with recombinant mouse (rm) DCC, rmNeogenin, and rmPUNC is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title