missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Nope/IGDCC4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1394-SP
This item is not returnable.
View return policy
Description
Nope/IGDCC4 Polyclonal specifically detects Nope/IGDCC4 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| Nope/IGDCC4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DDM36, hDDM36, IGDCC4, immunoglobulin superfamily DCC subclass member 4, immunoglobulin superfamily, DCC subclass, member 4, KIAA1628, likely ortholog of mouse neighbor of Punc E11, LOC57722, Neighbor of punc e11, NOPEFLJ42051, Protein DDM36 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 57722 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Q9EQS9 | |
| IGDCC4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Nope/IGDCC4, Gly22-His953, Accession # Q9EQS9 | |
| 25 μg | |
| Primary | |
| Detects mouse Nope/IGDCC4 in ELISAs and Western blots. In sandwich immunoassays, less than 0.3% cross-reactivity with recombinant mouse (rm) DCC, rmNeogenin, and rmPUNC is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction