missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NKp46/NCR1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
£167.00 - £399.00
Specifications
| Antigen | NKp46/NCR1 |
|---|---|
| Clone | 259018 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785518
![]() |
R&D Systems
MAB2225-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18199590
![]() |
R&D Systems
MAB2225 |
100 μg |
£399.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKp46/NCR1 Monoclonal specifically detects NKp46/NCR1 in Mouse samples. It is validated for Western Blot.Specifications
| NKp46/NCR1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8C567 | |
| 9437 | |
| Mouse myeloma cell line NS0-derived recombinant mouse NKp46 Glu22-Asn255 Accession # Q8C567 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse NKp46 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 10 - 20% cross-reactivity with recombinant human (rh) NKp46 and no cross-reactivity with rhNKp30 or rhNKp80. |
| 259018 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) | |
| NCR1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title