missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NKp46/NCR1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2225-SP
This item is not returnable.
View return policy
Description
NKp46/NCR1 Monoclonal specifically detects NKp46/NCR1 in Mouse samples. It is validated for Western Blot.
Specifications
| NKp46/NCR1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8C567 | |
| NCR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse NKp46 Glu22-Asn255 Accession # Q8C567 | |
| 25 μg | |
| Primary | |
| Detects mouse NKp46 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 10 - 20% cross-reactivity with recombinant human (rh) NKp46 and no cross-reactivity with rhNKp30 or rhNKp80. | |
| Mouse | |
| Purified |
| Western Blot | |
| 259018 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD335, CD335 antigen, hNKp46, Ly94, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, MAR-1, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NCR1, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9437 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction