missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NKG2A/CD159a APC-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
£168.00 - £506.00
Specifications
| Antigen | NKG2A/CD159a/KLRC1 |
|---|---|
| Clone | 705829 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18154491
|
R&D Systems
FAB6867A-025UG |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18145021
|
R&D Systems
FAB6867A-100UG |
100 μg |
£506.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKG2A/CD159a/KLRC1 Monoclonal antibody specifically detects NKG2A/CD159a/KLRC1 in Mouse samples. It is validated for Flow CytometrySpecifications
| NKG2A/CD159a/KLRC1 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| CD159 antigen-like family member A, CD159a, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, Klrc1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
| CHO Chinese hamster ovary cell line transfected with mouse NKG2A/CD159a, Accession # Q9Z202 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. |
| 705829 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| Mouse | |
| Q9Z202 | |
| 3821 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title