missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NKG2A/CD159a APC-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems FAB6867A-100UG
This item is not returnable.
View return policy
Description
NKG2A/CD159a/KLRC1 Monoclonal antibody specifically detects NKG2A/CD159a/KLRC1 in Mouse samples. It is validated for Flow Cytometry
Specifications
| NKG2A/CD159a/KLRC1 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| CD159 antigen-like family member A, CD159a, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, Klrc1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
| CHO Chinese hamster ovary cell line transfected with mouse NKG2A/CD159a, Accession # Q9Z202 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 705829 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9Z202 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3821 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction