missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Netrin-G2a Antibody, R&D Systems™

Goat Polyclonal Antibody
£146.00 - £398.00
Specifications
| Antigen | Netrin-G2a |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18754977
![]() |
R&D Systems
AF2744-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18011682
![]() |
R&D Systems
AF2744 |
100 μg |
£398.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Netrin-G2a Polyclonal specifically detects Netrin-G2a in Mouse samples. It is validated for Western Blot.Specifications
| Netrin-G2a | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| 84628 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Netrin-G2a Asp18-Arg504 Accession # NP_598007 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Netrin-G2a in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Netrin-1, rmNetrin-G1a, and rmNetrin-4 is observed and less than 1% cross-reactivity with recombinant chicken Netrin-2 is observed. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_598007 | |
| Ntng2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title