missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Netrin-G2a Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2744
This item is not returnable.
View return policy
Description
Netrin-G2a Polyclonal specifically detects Netrin-G2a in Mouse samples. It is validated for Western Blot.
Specifications
| Netrin-G2a | |
| Polyclonal | |
| Unconjugated | |
| NP_598007 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 84628 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Ntng2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Netrin-G2a Asp18-Arg504 Accession # NP_598007 | |
| 100 μg | |
| Primary | |
| Detects mouse Netrin-G2a in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Netrin-1, rmNetrin-G1a, and rmNetrin-4 is observed and less than 1% cross-reactivity with recombinant chicken Netrin-2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction