missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Nephronectin Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4298
This item is not returnable.
View return policy
Description
Nephronectin Polyclonal specifically detects Nephronectin in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| Nephronectin | |
| Polyclonal | |
| Unconjugated | |
| AAH68308 | |
| NPNT | |
| Mouse myeloma cell line NS0-derived recombinant mouse Nephronectin Asp20-Cys561 Accession # AAH68308 | |
| 100 μg | |
| Primary | |
| Detects mouse Nephronectin in direct ELISAs. | |
| Mouse | |
| IgG |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 1-15 ug/mL | |
| EGFL6L, Nctn, nephronectin, NPNT, POEM | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 255743 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction