missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Meteorin Antibody, R&D Systems™

Rat Monoclonal Antibody
£156.00 - £452.00
Specifications
| Antigen | Meteorin |
|---|---|
| Clone | 347511 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792789
![]() |
R&D Systems
MAB34752-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18121521
|
R&D Systems
MAB34752 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Meteorin Monoclonal specifically detects Meteorin in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Meteorin | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C16orf23, c380A1.2, METRN | |
| Metrn | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 347511 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8C1Q4 | |
| 79006 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Meteorin Gly22-Asp291 Accession # Q8C1Q4 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects mouse Meteorin in ELISAs.In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse CDNF, recombinant rat (rr) CNTF, or rrGDNF is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title