missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Meteorin Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB34752-SP
This item is not returnable.
View return policy
Description
Meteorin Monoclonal specifically detects Meteorin in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| Meteorin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C16orf23, c380A1.2, METRN | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 79006 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 347511 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q8C1Q4 | |
| Metrn | |
| Mouse myeloma cell line NS0-derived recombinant mouse Meteorin Gly22-Asp291 Accession # Q8C1Q4 | |
| 25 μg | |
| Primary | |
| Detects mouse Meteorin in ELISAs.In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse CDNF, recombinant rat (rr) CNTF, or rrGDNF is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction