missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Mer Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
£184.00 - £1775.00
Specifications
| Antigen | Mer |
|---|---|
| Clone | 108921 |
| Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18712789
![]() |
R&D Systems
MAB591-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18789597
![]() |
R&D Systems
MAB591-100 |
100 μg |
£452.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18734358
![]() |
R&D Systems
MAB591-500 |
500 μg |
£1775.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mer Monoclonal specifically detects Mer in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| Mer | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q60805 | |
| 10461 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Mer Glu23-Phe498 Accession # Q60805 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Mer in direct ELISAs and Western blots. Does not cross-react with recombinant human Mer, recombinant mouse (rm) Axl, or rmDtk. |
| 108921 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
| MERTK | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title