missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Mer Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB591-100
This item is not returnable.
View return policy
Description
Mer Monoclonal specifically detects Mer in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Mer | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10461 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 108921 | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q60805 | |
| MERTK | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Mer Glu23-Phe498 Accession # Q60805 | |
| 100 μg | |
| Primary | |
| Detects mouse Mer in direct ELISAs and Western blots. Does not cross-react with recombinant human Mer, recombinant mouse (rm) Axl, or rmDtk. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction