missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Matrilin-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
£168.00 - £353.00
Specifications
| Antigen | Matrilin-2 |
|---|---|
| Clone | 388207 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18708329
![]() |
R&D Systems
MAB3234-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18170701
![]() |
R&D Systems
MAB3234 |
100 μg |
£353.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Matrilin-2 Monoclonal specifically detects Matrilin-2 in Mouse samples. It is validated for Western Blot.Specifications
| Matrilin-2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| AAH05429 | |
| 4147 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Matrilin-2 Arg24-Arg956 Accession # AAH05429 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Matrilin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 20% cross-reactivity with recombinant human Matrilin-2 is observed and no cross-reactivity with recombinant mouse (rm) Matrilin-3 and rmMatrilin-4 is observed. |
| 388207 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MATN2, matrilin 2, matrilin-2, Matrilin2 | |
| MATN2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title