missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Matrilin-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3234-SP
This item is not returnable.
View return policy
Description
Matrilin-2 Monoclonal specifically detects Matrilin-2 in Mouse samples. It is validated for Western Blot.
Specifications
| Matrilin-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AAH05429 | |
| MATN2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Matrilin-2 Arg24-Arg956 Accession # AAH05429 | |
| 25 μg | |
| Primary | |
| Detects mouse Matrilin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 20% cross-reactivity with recombinant human Matrilin-2 is observed and no cross-reactivity with recombinant mouse (rm) Matrilin-3 and rmMatrilin-4 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 388207 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MATN2, matrilin 2, matrilin-2, Matrilin2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4147 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction