missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LOX-1/OLR1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
£184.00 - £510.00
Specifications
| Antigen | LOX-1/OLR1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18718667
![]() |
R&D Systems
AF1564-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18080262
|
R&D Systems
AF1564 |
100 μg |
£510.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LOX-1/OLR1 Polyclonal specifically detects LOX-1/OLR1 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
| LOX-1/OLR1 | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Polyclonal | |
| Goat | |
| Mouse | |
| CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1 | |
| OLR1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| AAG44998 | |
| 4973 | |
| Mouse myeloma cell line NS0-derived recombinant mouse LOX-1 (R&D Systems, Catalog # 1564-LX) Arg60-Ile363 Accession # AAG44998 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Selected for its ability to block receptor-ligand interaction in a functional ELISA assay. In Western blots, approximately 5% cross-reactivity with recombinant human LOX-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title