missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LOX-1/OLR1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF1564
This item is not returnable.
View return policy
Description
LOX-1/OLR1 Polyclonal specifically detects LOX-1/OLR1 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| LOX-1/OLR1 | |
| Polyclonal | |
| Unconjugated | |
| AAG44998 | |
| OLR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse LOX-1 (R&D Systems, Catalog # 1564-LX) Arg60-Ile363 Accession # AAG44998 | |
| 100 μg | |
| Primary | |
| Selected for its ability to block receptor-ligand interaction in a functional ELISA assay. In Western blots, approximately 5% cross-reactivity with recombinant human LOX-1 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4973 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction