missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Lipin 3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4064-SP
This item is not returnable.
View return policy
Description
Lipin 3 Polyclonal specifically detects Lipin 3 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Lipin 3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dJ450M14.2, dJ450M14.3, dJ620E11.2, EC 3.1.3.4, lipin 3, lipin 3-like, lipin-3, Lipin-3-like, LIPN3L, LPIN3, phosphatidate phosphatase LPIN3, SMP2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 64900 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q99PI4 | |
| LPIN3 | |
| E. coli-derived recombinant mouse Lipin 3 Thr428-Arg529 Accession # Q99PI4 | |
| 25 μg | |
| Primary | |
| Detects mouse Lipin 3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) Lipin 3 is observed and less than 5% cross-reactivity with rhLipin 1 and rhLipin 2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction