missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Lipin 3 Antibody, R&D Systems™

Goat Polyclonal Antibody
£156.00 - £398.00
Specifications
| Antigen | Lipin 3 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18736557
![]() |
R&D Systems
AF4064-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18099161
![]() |
R&D Systems
AF4064 |
100 μg |
£398.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lipin 3 Polyclonal specifically detects Lipin 3 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| Lipin 3 | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| dJ450M14.2, dJ450M14.3, dJ620E11.2, EC 3.1.3.4, lipin 3, lipin 3-like, lipin-3, Lipin-3-like, LIPN3L, LPIN3, phosphatidate phosphatase LPIN3, SMP2 | |
| LPIN3 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Q99PI4 | |
| 64900 | |
| E. coli-derived recombinant mouse Lipin 3 Thr428-Arg529 Accession # Q99PI4 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Lipin 3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) Lipin 3 is observed and less than 5% cross-reactivity with rhLipin 1 and rhLipin 2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title