Learn More
Invitrogen™ IL-9 Mouse Uncoated ELISA Kit
Specifically engineered for accurate and precise measurement of IL-9 protein levels from samples including serum, and cell culture supernatants. IL-9 is a 14 kd cytokine originally named P40 and identified by its proliferative effects on T cell population
Brand: Invitrogen™ 88-8092-76
Code : NEW
Additional Details : Weight : 0.47500kg
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
Coating Buffer. 10X PBS ELISA Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and standards.
Description
Mouse IL-9 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
32pg/mL | |
ELISA | |
20 x 96 Tests | |
Interleukin-9, T-cell growth factor P40 |
Mouse | |
IL-9 | |
2°C to 8°C except standard which should be stored at ≤-70°C | |
IL-9 |
For Research Use Only