missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-9 Mouse Uncoated ELISA Kit
Specifically engineered for accurate and precise measurement of IL-9 protein levels from samples including serum, and cell culture supernatants. IL-9 is a 14 kd cytokine originally named P40 and identified by its proliferative effects on T cell population
£174.00 - £546.00
Specifications
Target Species | Mouse |
---|---|
For Use With (Application) | ELISA |
Synonym | Interleukin-9, T-cell growth factor P40 |
Product Code | Brand | Quantity | Gene Symbol | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Gene Symbol | Price | Quantity & Availability | ||||
15571197
|
Invitrogen™
88-8092-22 |
2 x 96 Tests | CED,DPD1,LAP,TGFB,TGFbeta |
£174.00
Pack of 2 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
15511207
|
Invitrogen™
88-8092-88 |
10 x 96 Tests | IL-2 |
£368.00
Pack of 10 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||||
15501207
|
Invitrogen™
88-8092-86 |
10 x 96 Tests | IL-9 |
£392.00
Pack of 10 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||||
15591197
|
Invitrogen™
88-8092-77 |
20 x 96 Tests | IL-2 |
£524.00
Pack of 20 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||||
15581197
|
Invitrogen™
88-8092-76 |
20 x 96 Tests | IL-9 |
£546.00
Pack of 20 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||||
Description
Mouse IL-9 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
Mouse | |
ELISA | |
Interleukin-9, T-cell growth factor P40 |
For Research Use Only