missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-38/IL-1F10 Antibody, R&D Systems™

Rat Monoclonal Antibody
£203.00 - £1730.00
Specifications
| Antigen | IL-38/IL-1F10 |
|---|---|
| Clone | 798036R |
| Dilution | Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, ELISA |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18766729
![]() |
R&D Systems
MAB7774R-SP |
25 μg |
£203.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18735009
![]() |
R&D Systems
MAB7774R-100 |
100 μg |
£428.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18138800
|
R&D Systems
MAB7774R-01M |
1 mg |
£1460.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18150751
|
R&D Systems
MAB7774R-500 |
500 ug |
£1730.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| IL-38/IL-1F10 | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q8R459 | |
| 84639 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Detects mouse IL-38/IL-1F10 in direct ELISAs and Western blots. |
| 798036R | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 | |
| Il1f10 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title