missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-38/IL-1F10 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB7774R-01M
This item is not returnable.
View return policy
Description
IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| IL-38/IL-1F10 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 84639 | |
| Mouse | |
| Purified |
| ELISA, Flow Cytometry, Western Blot | |
| 798036R | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, ELISA | |
| Q8R459 | |
| Il1f10 | |
| E.coli-derived recombinant mouse IL-38/IL-1F10 Met1-Arg152 Accession No. Q8R459 | |
| 1 mg | |
| Primary | |
| Detects mouse IL-38/IL-1F10 in direct ELISAs and Western blots. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction