missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-36 alpha/IL-1F6 Antibody, R&D Systems™

Rat Monoclonal Antibody
£185.00 - £484.00
Specifications
| Antigen | IL-36 alpha/IL-1F6 |
|---|---|
| Clone | 275302 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18711717
![]() |
R&D Systems
MAB22972-SP |
25 μg |
£185.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18740047
|
R&D Systems
MAB22972-100 |
100 μg |
£484.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-36 alpha/IL-1F6 Monoclonal antibody specifically detects IL-36 alpha/IL-1F6 in Mouse samples. It is validated for ELISASpecifications
| IL-36 alpha/IL-1F6 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha | |
| E. coli-derived recombiMouset mouse IL-36 alpha/IL-1F6, Met1-His160, Accession # Q9JLA2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 275302 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9JLA2 | |
| 27179 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title