Learn More
Mouse IL-36 alpha/IL-1F6 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand:  R&D Systems MAB22972-100
 View more versions of this product
Product Code. 18740047
  • £484.00 / 100µg
Description
IL-36 alpha/IL-1F6 Monoclonal antibody specifically detects IL-36 alpha/IL-1F6 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-36 alpha/IL-1F6
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha
E. coli-derived recombiMouset mouse IL-36 alpha/IL-1F6, Met1-His160, Accession # Q9JLA2
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2a
ELISA
275302
ELISA
Q9JLA2
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
27179
Mouse
Purified