missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-1ra/IL-1F3 Antibody, R&D Systems™

Rat Monoclonal Antibody
£177.00 - £397.00
Specifications
| Antigen | IL-1ra/IL-1F3/IL1RN |
|---|---|
| Clone | 962802 |
| Dilution | Immunocytochemistry 3-25 μg;/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18714507
![]() |
R&D Systems
MAB4802-SP |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18704407
![]() |
R&D Systems
MAB4802-100 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Mouse samples. It is validated for Immunocytochemistry.Specifications
| IL-1ra/IL-1F3/IL1RN | |
| Immunocytochemistry 3-25 μg;/mL | |
| Monoclonal | |
| Rat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DIRA, ICIL-1RA, IL1F3, IL-1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
| E. coli-derived recombinant mouse IL-1ra/IL-1F3, Arg27-Gln178, Accession No. P258085 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IL-1ra/IL-1F3 in direct ELISAs. |
| 962802 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| P258085 | |
| 3557.0 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title