missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-1ra/IL-1F3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4802-100
This item is not returnable.
View return policy
Description
IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Mouse samples. It is validated for Immunocytochemistry.
Specifications
| IL-1ra/IL-1F3/IL1RN | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DIRA, ICIL-1RA, IL1F3, IL-1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
| E. coli-derived recombinant mouse IL-1ra/IL-1F3, Arg27-Gln178, Accession No. P258085 | |
| 100 μg | |
| 3557.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry | |
| 962802 | |
| Immunocytochemistry 3-25 μg;/mL | |
| P258085 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse IL-1ra/IL-1F3 in direct ELISAs. | |
| Mouse | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction