missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-17AF (heterodimer) Mouse Uncoated ELISA Kit

Specifically engineered for accurate and precise measurement protein levels from samples including serum and cell culture supernatants.

Brand:  Invitrogen™ 88-8711-88

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15521237

  • £368.00 / Pack of 10
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Alert:

Contains Sodium azide


Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer 10X PBS ELISA Coating Buffer.
Assay Diluent. 5X concentrated
Sample Diluent A
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

Description

Description

Interleukin-17A (IL-17A) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. The cDNA encoding human IL-17A was isolated from a library of CD4+ T cells; the encoded protein exhibits 72 percent amino acid identity with HVS13 , an open reading frame from a T lymphotropic Herpesvirus saimiri, and 63 percent with mouse CTLA-8 (cytotoxic T-lymphocyte associated antigen-8). Human IL-17A exists as glycosylated 20-30 kD homodimers. High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells. IL-17A enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. Human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A binds to a receptor that binds also to HVS13 (viral IL-17) and to CTLA-8.This recombinant protein is E. coli expressed. Insect cell expressed Human IL-17A Recombinant Protein (cat. 14-8180) is also available.
Specifications

Specifications

Q62386,Q7TNI7
16 pg/mL
ELISA Kit
Mouse
Colorimetric Microplate Reader
GCP-1,GCP1,IL8,LECT,LUCT,LYNAP,MDNCF,MONAP,NAF,NAP-1,NAP1
HRP
RUO
2°C to 8°C
Mouse
24 hr. 30 min.
16-2,000 pg/mL
Biotin
Serum, Supernatant
ELISA
16171, 257630
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, Coating Buffer 10X PBS ELISA Coating Buffer., Assay Diluent: 5X concentrated, Sample Diluent A: 12 mL of a 1X solution per plate, Detection enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -44; -76; -86)
10 x 96 Tests
Serum, 50 μL; Supernatant, 50 μL
Interleukin-17A/F
1 hr. 45 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only