missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-17AF (heterodimer) Mouse Uncoated ELISA Kit

Specifically engineered for accurate and precise measurement protein levels from samples including serum and cell culture supernatants.

£145.00 - £460.00

Specifications

For Use With (Application) ELISA
Synonym Interleukin-17A/F
Target Mouse
Sensitivity 16 pg/mL
Storage Requirements 2°C to 8°C
View More Specs

Alert:

Contains Sodium azide


Products 5
Product Code Brand Quantity Gene Symbols Price Quantity & Availability  
Product Code Brand Quantity Gene Symbols Price Quantity & Availability  
15511237 View Documents Invitrogen™
88-8711-86
10 x 96 Tests £322.00
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Add to basket
 
15521237 View Documents Invitrogen™
88-8711-88
10 x 96 Tests GCP-1,GCP1,IL8,LECT,LUCT,LYNAP,MDNCF,MONAP,NAF,NAP-1,NAP1 £301.00
Pack of 10 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Add to basket
 
15581227 View Documents Invitrogen™
88-8711-22
2 x 96 Tests Ctla-8,Ctla8,IL-17,IL-17A,Il17 £145.00
Pack of 2 View alternative products
Save Up to 
»
EU Stock 5
Add to basket
 
15591227 View Documents Invitrogen™
88-8711-76
20 x 96 Tests £460.00
Pack of 20 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Add to basket
 
15501237 View Documents Invitrogen™
88-8711-77
20 x 96 Tests £434.00
Pack of 20 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Add to basket
 
Description

Description

  • Mouse IL-17AF (heterodimer) ELISA kit contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
  • By incorporating a monoclonal antibody to IL-17A as the capture reagent and IL-17AF as the detection reagent, this second generation kit has increased sensitivity with a range from 16 to 2000pg/mL and no cross-reactivity to the IL-17A or IL-17F homodimers
Specifications

Specifications

ELISA
Mouse
2°C to 8°C
Interleukin-17A/F
16 pg/mL
Mouse

For Research Use Only