missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-17 Quantikine ELISA Kit
Description
The Mouse IL-17 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-17/IL-17A in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
Specifications
Specifications
| Assay Range | 10.90 - 700pg/mL |
| Assay Sensitivity | 5pg/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
| Specificity | Natural and Recombinant Mouse IL-17 |
| Sufficient For | 6 x 96-well Plates |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?