missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG Antibody Pair, Novus Biologicals™

Antibody Pair

£351.00 - £696.00

Specifications

Antigen Mouse IgG
Applications ELISA
Conjugate Unconjugated
Regulatory Status RUO
Target Species Mouse
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30609580
View Documents
Novus Biologicals
NBP3-48426-15Plates
15 Plates
£696.00
1 set
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30611886
View Documents
Novus Biologicals
NBP3-48426-5Plates
5 Plates
£351.00
1 set
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mouse IgG nan antibody specifically detects Mouse IgG in Mouse samples. It is validated for ELISA
Specifications

Specifications

Mouse IgG
Unconjugated
Mouse
Storage of components varies. See protocol for specific instructions.
ELISA
RUO
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.