missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG Antibody Pair, Novus Biologicals™

Product Code. 30609580 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30609580 15 Plates 1 set
30611886 5 Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30609580 Supplier Novus Biologicals Supplier No. NBP34842615Plates
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibody Pair

Mouse IgG nan antibody specifically detects Mouse IgG in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG
Applications ELISA
Conjugate Unconjugated
Quantity 15 Plates
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.