missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IGFBP-3 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
£168.00 - £487.00
Specifications
| Antigen | IGFBP-3 |
|---|---|
| Clone | 138202 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18778968
![]() |
R&D Systems
MAB775-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18170741
|
R&D Systems
MAB775 |
500 μg |
£487.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGFBP-3 Monoclonal specifically detects IGFBP-3 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| IGFBP-3 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 | |
| IGFBP3 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 138202 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P47878 | |
| 3486 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-3 Pro22-Gln291 (Arg250Gln, Gln259Arg, and Arg271Pro) Accession # P47878 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse IGFBP-3 in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human IGFBP-1, rhIGFBP-2, rhIGFBP-3, rhIGFBP-4, rhIGFBP-5, rhIGFBP-6, recombinant mouse (rm) IGFBP-2, rmIGFBP-5, or rmIGFBP-6. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title