missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IGFBP-3 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB775-SP
This item is not returnable.
View return policy
Description
IGFBP-3 Monoclonal specifically detects IGFBP-3 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| IGFBP-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3486 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, ELISA | |
| 138202 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P47878 | |
| IGFBP3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-3 Pro22-Gln291 (Arg250Gln, Gln259Arg, and Arg271Pro) Accession # P47878 | |
| 25 μg | |
| Primary | |
| Detects mouse IGFBP-3 in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human IGFBP-1, rhIGFBP-2, rhIGFBP-3, rhIGFBP-4, rhIGFBP-5, rhIGFBP-6, recombinant mouse (rm) IGFBP-2, rmIGFBP-5, or rmIGFBP-6. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction