missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse HRP-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
£146.00 - £452.00
Specifications
| Antigen | HRP-1/HDGFL1 |
|---|---|
| Clone | 249308 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713427
![]() |
R&D Systems
MAB1868-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18190721
![]() |
R&D Systems
MAB1868 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HRP-1/HDGFL1 Monoclonal specifically detects HRP-1/HDGFL1 in Mouse samples. It is validated for Western Blot.Specifications
| HRP-1/HDGFL1 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dJ309H15.1, HDGF (hepatoma-derived growth factor) like, hepatoma derived growth factor-like 1, hepatoma-derived growth factor-like protein 1, HRP1, PWWP domain containing 1, PWWP domain-containing protein 1, PWWP1 | |
| HDGFL1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 249308 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| NP_032258 | |
| 154150 | |
| E. coli-derived recombinant human HRP-1 Met1-Leu283 Accession # NP_032258 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse HRP-1 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title