missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse HRP-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1868-SP
This item is not returnable.
View return policy
Description
HRP-1/HDGFL1 Monoclonal specifically detects HRP-1/HDGFL1 in Mouse samples. It is validated for Western Blot.
Specifications
| HRP-1/HDGFL1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dJ309H15.1, HDGF (hepatoma-derived growth factor) like, hepatoma derived growth factor-like 1, hepatoma-derived growth factor-like protein 1, HRP1, PWWP domain containing 1, PWWP domain-containing protein 1, PWWP1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 154150 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 249308 | |
| Western Blot 1 ug/mL | |
| NP_032258 | |
| HDGFL1 | |
| E. coli-derived recombinant human HRP-1 Met1-Leu283 Accession # NP_032258 | |
| 25 μg | |
| Primary | |
| Detects mouse HRP-1 in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction