missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse HPRG Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB1905
Additional Details : Weight : 0.00970kg
Description
HPRG Monoclonal specifically detects HPRG in Mouse samples. It is validated for Western Blot.Specifications
HPRG | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
BAB33094 | |
HRG | |
Mouse myeloma cell line NS0-derived recombinant mouse HPRG Leu19-Lys525 Accession # BAB33094 | |
500 μg | |
Primary | |
Detects mouse HPRG in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 20% cross-reactivity with recombinant human (rh) HPRG is observed and no cross-reactivity with rhCystatin D, F, S, SA, SN, recombinant mouse (rm) Cystatin B, C, E/M, rmKininogen, and rhKininostatin is observed. | |
Mouse | |
Purified |
Western Blot | |
242425 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp779H1622, Histidine-proline-rich glycoprotein, histidine-rich glycoprotein, HPRGHRGP, HRG | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3273 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
For Research Use Only