missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse HPRG Antibody, R&D Systems™

Rat Monoclonal Antibody
£167.00 - £478.00
Specifications
| Antigen | HPRG |
|---|---|
| Clone | 242425 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18731927
![]() |
R&D Systems
MAB1905-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18119200
![]() |
R&D Systems
MAB1905 |
500 μg |
£478.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HPRG Monoclonal specifically detects HPRG in Mouse samples. It is validated for Western Blot.Specifications
| HPRG | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| BAB33094 | |
| 3273 | |
| Mouse myeloma cell line NS0-derived recombinant mouse HPRG Leu19-Lys525 Accession # BAB33094 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse HPRG in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 20% cross-reactivity with recombinant human (rh) HPRG is observed and no cross-reactivity with rhCystatin D, F, S, SA, SN, recombinant mouse (rm) Cystatin B, C, E/M, rmKininogen, and rhKininostatin is observed. |
| 242425 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp779H1622, Histidine-proline-rich glycoprotein, histidine-rich glycoprotein, HPRGHRGP, HRG | |
| HRG | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title