missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Hip Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
£167.00 - £460.00
Especificaciones
| Antigen | Hip |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
18718967
![]() |
R&D Systems
AF1568-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18012922
|
R&D Systems
AF1568 |
100 μg |
£460.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
Hip Polyclonal specifically detects Hip in Mouse samples. It is validated for Western Blot.Especificaciones
| Hip | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| FLJ90230, hedgehog interacting protein, hedgehog-interacting protein, HHIP, HIPFLJ20992 | |
| HHIP | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q7TN16 | |
| 64399 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Hip Lys24-Arg678 (Asn29Ser) Accession # Q7TN16 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Hip in direct ELISAs and Western blots. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto