missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Hip Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF1568
This item is not returnable.
View return policy
Description
Hip Polyclonal specifically detects Hip in Mouse samples. It is validated for Western Blot.
Specifications
| Hip | |
| Polyclonal | |
| Unconjugated | |
| Q7TN16 | |
| HHIP | |
| Mouse myeloma cell line NS0-derived recombinant mouse Hip Lys24-Arg678 (Asn29Ser) Accession # Q7TN16 | |
| 100 μg | |
| Primary | |
| Detects mouse Hip in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| FLJ90230, hedgehog interacting protein, hedgehog-interacting protein, HHIP, HIPFLJ20992 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 64399 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction