missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse H60 Antibody, R&D Systems™

Goat Polyclonal Antibody
£171.00 - £442.00
Specifications
| Antigen | H60 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction |
| Applications | Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18766107
![]() |
R&D Systems
AF1155-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18038731
![]() |
R&D Systems
AF1155 |
100 μg |
£442.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
H60 Polyclonal specifically detects H60 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction.Specifications
| H60 | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction | |
| Polyclonal | |
| Goat | |
| Mouse | |
| 15101 | |
| Mouse myeloma cell line NS0-derived recombinant mouse H60 Asp30-Gln212 Accession # Q3TDZ7 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse H60 in direct ELISAs and Western blots. |
| LYOPH | |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| Q3TDZ7 | |
| H60a | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title