missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse H60 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1155
This item is not returnable.
View return policy
Description
H60 Polyclonal specifically detects H60 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction.
Specifications
| H60 | |
| Polyclonal | |
| Unconjugated | |
| Q3TDZ7 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 15101 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction | |
| H60a | |
| Mouse myeloma cell line NS0-derived recombinant mouse H60 Asp30-Gln212 Accession # Q3TDZ7 | |
| 100 μg | |
| Primary | |
| Detects mouse H60 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction