missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GDF-7/BMP-12 Antibody, R&D Systems™

Goat Polyclonal Antibody
£147.00 - £413.00
Specifications
| Antigen | GDF-7/BMP-12 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18755817
![]() |
R&D Systems
AF779-SP |
25 μg |
£147.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18053142
|
R&D Systems
AF779 |
100 μg |
£413.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-7/BMP-12 Polyclonal specifically detects GDF-7/BMP-12 in Mouse samples. It is validated for Western Blot.Specifications
| GDF-7/BMP-12 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| BMP12, BMP-12, GDF7, GDF-7, growth differentiation factor 7, growth/differentiation factor 7 | |
| GDF7 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P43029 | |
| 151449 | |
| E. coli-derived recombinant mouse GDF-7 (R&D Systems, Catalog # 779-G7) Thr316-Arg461 Accession # P43029 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse GDF-7 in direct ELISAs and Western blots. In these formats, approximately 10% cross-reactivity with recombinant mouse (rm) GDF-5 and rmGDF-6 is observed and less than 1% cross-reactivity with rmGDF-1, rmGDF-3, rmGDF-8, rmGDF-9, and recombinant human GDF-11 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title