missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GDF-7/BMP-12 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF779
This item is not returnable.
View return policy
Description
GDF-7/BMP-12 Polyclonal specifically detects GDF-7/BMP-12 in Mouse samples. It is validated for Western Blot.
Specifications
| GDF-7/BMP-12 | |
| Polyclonal | |
| Unconjugated | |
| P43029 | |
| GDF7 | |
| E. coli-derived recombinant mouse GDF-7 (R&D Systems, Catalog # 779-G7) Thr316-Arg461 Accession # P43029 | |
| 100 μg | |
| Primary | |
| Detects mouse GDF-7 in direct ELISAs and Western blots. In these formats, approximately 10% cross-reactivity with recombinant mouse (rm) GDF-5 and rmGDF-6 is observed and less than 1% cross-reactivity with rmGDF-1, rmGDF-3, rmGDF-8, rmGDF-9, and recombinant human GDF-11 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| BMP12, BMP-12, GDF7, GDF-7, growth differentiation factor 7, growth/differentiation factor 7 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 151449 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction